siRNA Details
Gene Symbol: Vwa8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008770921.1 | XP_008769143.1 | 5146 | |||
GenBank mRNA | BC083871.1 | - | 6 | 820 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Vwa8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008770921.1 | XP_008769143.1 | 5146 | |||
GenBank mRNA | BC083871.1 | - | 6 | 820 |