siRNA Details
| Gene Symbol: Trim52 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106056.1 | NP_001099526.1 | 3 | 775 | ||
| XM_006252435.2 | XP_006252497.1 | 1 | 1666 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Trim52| Gene Symbol: Trim52 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106056.1 | NP_001099526.1 | 3 | 775 | ||
| XM_006252435.2 | XP_006252497.1 | 1 | 1666 | |||