siRNA Details
| Gene Symbol: Tnnc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034105.1 | NP_001029277.1 | 3 | 72 | ||
| XM_008770987.1 | XP_008769209.1 | 4 | 216 | |||
| GenBank mRNA | DQ062205.1 | - | 3 | 72 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tnnc1| Gene Symbol: Tnnc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034105.1 | NP_001029277.1 | 3 | 72 | ||
| XM_008770987.1 | XP_008769209.1 | 4 | 216 | |||
| GenBank mRNA | DQ062205.1 | - | 3 | 72 | ||