siRNA Details
Gene Symbol: Zfp488 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006252711.2 | XP_006252773.1 | 2 | 1208 | ||
XM_006252712.2 | XP_006252774.1 | 2 | 1142 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Zfp488 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006252711.2 | XP_006252773.1 | 2 | 1208 | ||
XM_006252712.2 | XP_006252774.1 | 2 | 1142 |