siRNA Details
| Gene Symbol: Haus8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024971.2 | NP_001020142.2 | 2 | 232 | ||
| XM_008771074.1 | XP_008769296.1 | 2 | 281 | |||
| XM_008771075.1 | XP_008769297.1 | 2 | 281 | |||
| XM_008771076.1 | XP_008769298.1 | 2 | 281 | |||
| XM_008771077.1 | XP_008769299.1 | 2 | 280 | |||
| GenBank mRNA | BC091198.1 | - | 2 | 221 | 7376974 | |
| CB691625.1 | - | 2 | 233 | |||
| DY317896.1 | - | 2 | 192 | |||
| FQ225627.1 | - | 2 | 244 | |||

Haus8
Genomic Map