siRNA Details
| Gene Symbol: Ushbp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033687.1 | NP_001028859.1 | 4 | 467 | ||
| GenBank mRNA | BC101905.1 | - | 4 | 467 | 7934347 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ushbp1| Gene Symbol: Ushbp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033687.1 | NP_001028859.1 | 4 | 467 | ||
| GenBank mRNA | BC101905.1 | - | 4 | 467 | 7934347 | |