siRNA Details
| Gene Symbol: Babam1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001006964.1 | NP_001006965.1 | 7 | 797 | ||
| XM_006252857.2 | XP_006252919.1 | 7 | 821 | |||
| GenBank mRNA | BC083685.1 | - | 7 | 797 | 7189038 | |
| FQ214886.1 | - | 7 | 788 | |||
| FQ233705.1 | - | 7 | 808 | |||

Babam1
Genomic Map