siRNA Details
Gene Symbol: Colgalt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106067.2 | NP_001099537.1 | 5 | 731 | ||
GenBank mRNA | BC160899.1 | - | 5 | 731 | 7453718 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Colgalt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106067.2 | NP_001099537.1 | 5 | 731 | ||
GenBank mRNA | BC160899.1 | - | 5 | 731 | 7453718 |