siRNA Details
Gene Symbol: Ccdc124 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106071.1 | NP_001099541.1 | 5 | 984 | ||
XM_006252867.2 | XP_006252929.1 | 5 | 620 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Ccdc124 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106071.1 | NP_001099541.1 | 5 | 984 | ||
XM_006252867.2 | XP_006252929.1 | 5 | 620 |