siRNA Details
| Gene Symbol: Dctd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013882.2 | NP_001013904.2 | 3, 4 | 445 | ||
| GenBank mRNA | BC087138.1 | - | 3, 4 | 400 | 7125992 | |
| DY317307.1 | - | 3, 4 | 413 | |||
| EV776012.1 | - | 3, 4 | 470 | |||

Dctd
Genomic Map