siRNA Details
| Gene Symbol: Cldn23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033062.1 | NP_001028234.1 | 1 | 631 | ||
| GenBank mRNA | BC100630.1 | - | 1 | 631 | 7938080 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cldn23| Gene Symbol: Cldn23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033062.1 | NP_001028234.1 | 1 | 631 | ||
| GenBank mRNA | BC100630.1 | - | 1 | 631 | 7938080 | |