siRNA Details
| Gene Symbol: Tti2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013883.1 | NP_001013905.1 | 1 | 405 | ||
| GenBank mRNA | BC082008.1 | - | 1 | 405 | 7131694 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tti2| Gene Symbol: Tti2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013883.1 | NP_001013905.1 | 1 | 405 | ||
| GenBank mRNA | BC082008.1 | - | 1 | 405 | 7131694 | |