siRNA Details
| Gene Symbol: Catsper3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106101.1 | NP_001099571.1 | 6 | 860 | ||
| XM_006253572.2 | XP_006253634.1 | 6 | 2099 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Catsper3| Gene Symbol: Catsper3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106101.1 | NP_001099571.1 | 6 | 860 | ||
| XM_006253572.2 | XP_006253634.1 | 6 | 2099 | |||