siRNA Details
| Gene Symbol: Gadd45g | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001077640.1 | NP_001071108.1 | 1 | 116 | ||
| GenBank mRNA | AB020978.1 | - | 1 | 116 | ||
| BC168643.1 | - | 1 | 116 | 9092341 | ||
| FQ218856.1 | - | 1 | 134 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gadd45g| Gene Symbol: Gadd45g | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001077640.1 | NP_001071108.1 | 1 | 116 | ||
| GenBank mRNA | AB020978.1 | - | 1 | 116 | ||
| BC168643.1 | - | 1 | 116 | 9092341 | ||
| FQ218856.1 | - | 1 | 134 | |||