siRNA Details
| Gene Symbol: Diras2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001169578.1 | NP_001163049.1 | 1 | 194 | ||
| XM_006253671.2 | XP_006253733.1 | 2 | 259 | |||
| XM_008771489.1 | XP_008769711.1 | 2 | 278 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Diras2| Gene Symbol: Diras2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001169578.1 | NP_001163049.1 | 1 | 194 | ||
| XM_006253671.2 | XP_006253733.1 | 2 | 259 | |||
| XM_008771489.1 | XP_008769711.1 | 2 | 278 | |||