siRNA Details
| Gene Symbol: Gmds | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039606.1 | NP_001034695.1 | 5 | 489 | ||
| GenBank mRNA | BC104708.1 | - | 5 | 489 | 7317156 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gmds| Gene Symbol: Gmds | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039606.1 | NP_001034695.1 | 5 | 489 | ||
| GenBank mRNA | BC104708.1 | - | 5 | 489 | 7317156 | |