siRNA Details
| Gene Symbol: Acot13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106111.1 | NP_001099581.1 | 1 | 267 | ||
| GenBank mRNA | FQ217526.1 | - | 1 | 217 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Acot13| Gene Symbol: Acot13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106111.1 | NP_001099581.1 | 1 | 267 | ||
| GenBank mRNA | FQ217526.1 | - | 1 | 217 | ||