siRNA Details
Gene Symbol: Epdr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007625.1 | NP_001007626.1 | 1, 2 | 488 | ||
GenBank mRNA | BC083701.1 | - | 1, 2 | 488 | 7190104 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Epdr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007625.1 | NP_001007626.1 | 1, 2 | 488 | ||
GenBank mRNA | BC083701.1 | - | 1, 2 | 488 | 7190104 |