siRNA Details
| Gene Symbol: Hax1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181627.2 | NP_853658.1 | 379 | |||
| GenBank mRNA | AY291062.1 | - | 379 | |||
| AY291063.2 | - | 574 | ||||
| AY919343.1 | - | 358 | ||||
| FQ213378.1 | - | 396 | ||||
| FQ213584.1 | - | 396 | ||||
| FQ214037.1 | - | 397 | ||||
| FQ214498.1 | - | 407 | ||||
| FQ229190.1 | - | 396 | ||||
| FQ232927.1 | - | 396 | ||||

Hax1
Genomic Map