siRNA Details
| Gene Symbol: Mrpl32 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106116.1 | NP_001099586.1 | 3 | 558 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mrpl32| Gene Symbol: Mrpl32 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106116.1 | NP_001099586.1 | 3 | 558 | ||