siRNA Details
| Gene Symbol: Fbxo18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106119.1 | NP_001099589.1 | 16, 17 | 2528 | ||
| XM_006254200.2 | XP_006254262.1 | 16, 17 | 2629 | |||
| XM_006254201.2 | XP_006254263.1 | 16, 17 | 2655 | |||
| XM_006254202.2 | XP_006254264.1 | 16, 17 | 2558 | |||
| XM_006254204.2 | XP_006254266.1 | 16, 17 | 2476 | |||

Fbxo18
Genomic Map