siRNA Details
| Gene Symbol: Fam188a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106122.1 | NP_001099592.1 | 12, 13 | 1200 | ||
| GenBank mRNA | FQ214325.1 | - | 9, 10 | 871 | ||
| FQ214453.1 | - | 7, 8 | 730 | |||
| FQ230167.1 | - | 6, 7 | 559 | |||

Fam188a
Genomic Map