siRNA Details
| Gene Symbol: Fam188a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106122.1 | NP_001099592.1 | 13 | 1251 | ||
| GenBank mRNA | FQ214325.1 | - | 10 | 922 | ||
| FQ214453.1 | - | 8 | 781 | |||
| FQ230167.1 | - | 7 | 610 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fam188a| Gene Symbol: Fam188a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106122.1 | NP_001099592.1 | 13 | 1251 | ||
| GenBank mRNA | FQ214325.1 | - | 10 | 922 | ||
| FQ214453.1 | - | 8 | 781 | |||
| FQ230167.1 | - | 7 | 610 | |||