siRNA Details
Gene Symbol: Spag6l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106125.1 | NP_001099595.1 | 4 | 301 | ||
XM_006254307.2 | XP_006254369.1 | 5 | 4215 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Spag6l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106125.1 | NP_001099595.1 | 4 | 301 | ||
XM_006254307.2 | XP_006254369.1 | 5 | 4215 |