siRNA Details
| Gene Symbol: Zfp236 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006255011.2 | XP_006255073.1 | 30 | 5453 | ||
| XM_006255013.2 | XP_006255075.1 | 29 | 5273 | |||
| XM_008772218.1 | XP_008770440.1 | 30 | 5587 | |||
| XM_008772219.1 | XP_008770441.1 | 29 | 5321 | |||

Zfp236
Genomic Map