siRNA Details
| Gene Symbol: Matr3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019149.3 | NP_062022.2 | 12 | 2278 | ||
| XM_006254534.2 | XP_006254596.1 | 12 | 2300 | |||
| XM_006254535.2 | XP_006254597.1 | 12 | 2251 | |||
| GenBank mRNA | AB205483.1 | - | 11 | 3048 | ||
| M63485.1 | - | 12 | 2263 | |||

Matr3
Genomic Map