siRNA Details
| Gene Symbol: Matr3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019149.3 | NP_062022.2 | 15 | 2764 | ||
| XM_006254534.2 | XP_006254596.1 | 15 | 2786 | |||
| XM_006254535.2 | XP_006254597.1 | 14 | 2615 | |||
| GenBank mRNA | AB205483.1 | - | 14 | 3534 | ||
| FQ233097.1 | - | 3 | 214 | |||

Matr3
Genomic Map