siRNA Details
Gene Symbol: Fbln5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019153.3 | NP_062026.2 | 5 | 644 | ||
XM_006240459.2 | XP_006240521.1 | 5 | 875 | |||
GenBank mRNA | AF112153.1 | - | 5 | 768 | ||
AF137350.1 | - | 5 | 571 | |||
BC078845.1 | - | 5 | 644 | 7123752 |
