siRNA Details
| Gene Symbol: Spry4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106150.1 | NP_001099620.1 | 2 | 543 | ||
| XM_008772067.1 | XP_008770289.1 | 2 | 1400 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Spry4| Gene Symbol: Spry4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106150.1 | NP_001099620.1 | 2 | 543 | ||
| XM_008772067.1 | XP_008770289.1 | 2 | 1400 | |||