siRNA Details
| Gene Symbol: Ndufa2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106153.1 | NP_001099623.1 | 1, 2 | 131 | ||
| GenBank mRNA | FQ212326.1 | - | 1, 2 | 107 | ||
| FQ216914.1 | - | 1, 2 | 109 | |||
| FQ216957.1 | - | 1, 2 | 126 | |||
| FQ217727.1 | - | 1, 2 | 105 | |||
| FQ223951.1 | - | 1, 2 | 106 | |||
| FQ224026.1 | - | 1, 2 | 109 | |||

Ndufa2
Genomic Map