siRNA Details
Gene Symbol: Spata24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025636.2 | NP_001020807.2 | 4 | 336 | ||
XM_006254545.2 | XP_006254607.1 | 4 | 455 | |||
GenBank mRNA | BQ207964.1 | - | 1 | 322 | ||
DQ080016.1 | - | 4 | 336 |
