siRNA Details
| Gene Symbol: Spata24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025636.2 | NP_001020807.2 | 4 | 505 | ||
| GenBank mRNA | BQ207964.1 | - | 1 | 153 | ||
| CR463891.1 | - | 1 | 163 | |||
| DQ080016.1 | - | 4 | 505 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Spata24| Gene Symbol: Spata24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025636.2 | NP_001020807.2 | 4 | 505 | ||
| GenBank mRNA | BQ207964.1 | - | 1 | 153 | ||
| CR463891.1 | - | 1 | 163 | |||
| DQ080016.1 | - | 4 | 505 | |||