siRNA Details
| Gene Symbol: Srp19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106157.2 | NP_001099627.1 | 4 | 255 | ||
| GenBank mRNA | BC167043.1 | - | 4 | 255 | 7319792 | |
| FQ233350.1 | - | 4 | 305 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Srp19| Gene Symbol: Srp19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106157.2 | NP_001099627.1 | 4 | 255 | ||
| GenBank mRNA | BC167043.1 | - | 4 | 255 | 7319792 | |
| FQ233350.1 | - | 4 | 305 | |||