siRNA Details
Gene Symbol: Srp19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106157.2 | NP_001099627.1 | 5 | 342 | ||
GenBank mRNA | BC167043.1 | - | 5 | 342 | 7319792 | |
FQ233350.1 | - | 5 | 392 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Srp19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106157.2 | NP_001099627.1 | 5 | 342 | ||
GenBank mRNA | BC167043.1 | - | 5 | 342 | 7319792 | |
FQ233350.1 | - | 5 | 392 |