siRNA Details
Gene Symbol: Brd8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008509.1 | NP_001008509.1 | 8 | 908 | ||
XM_006254562.2 | XP_006254624.1 | 9 | 1094 | |||
XM_006254563.2 | XP_006254625.1 | 9 | 1095 | |||
XM_008772029.1 | XP_008770251.1 | 9 | 1132 | |||
GenBank mRNA | AB180485.1 | - | 8 | 908 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_030147, AK132365, AK032320, AK042446, BC025644.
Gene Ontology Categories:
