siRNA Details
| Gene Symbol: Vtn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019156.2 | NP_062029.2 | 5 | 882 | ||
| GenBank mRNA | AY318963.1 | - | 870 | |||
| BC105821.1 | - | 5 | 882 | 7371104 | ||
| FQ209762.1 | - | 1 | 56 | |||
| FQ210146.1 | - | 3 | 502 | |||
| FQ210518.1 | - | 5 | 887 | |||
| FQ218401.1 | - | 3 | 455 | |||
| U44845.1 | - | 5 | 872 | |||

Vtn
Genomic Map