siRNA Details
Gene Symbol: Dsg3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191079.1 | NP_001178008.1 | 12 | 1689 | ||
XM_008771934.1 | XP_008770156.1 | 12 | 1689 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Dsg3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191079.1 | NP_001178008.1 | 12 | 1689 | ||
XM_008771934.1 | XP_008770156.1 | 12 | 1689 |