siRNA Details
Gene Symbol: Syn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034020.1 | NP_001029192.1 | 10 | 1303 | ||
NM_019159.1 | NP_062032.1 | 10 | 1303 | |||
GenBank mRNA | M27925.1 | - | 10 | 1303 | ||
M27926.1 | - | 10 | 1303 |
