siRNA Details
| Gene Symbol: Uts2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019160.2 | NP_062033.1 | 4 | 348 | ||
| GenBank mRNA | AF172174.1 | - | 4 | 348 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Uts2| Gene Symbol: Uts2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019160.2 | NP_062033.1 | 4 | 348 | ||
| GenBank mRNA | AF172174.1 | - | 4 | 348 | ||