siRNA Details
| Gene Symbol: Cmtm3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106164.1 | NP_001099634.1 | 2 | 285 | ||
| XM_006255049.2 | XP_006255111.1 | 3 | 368 | |||
| XM_008772241.1 | XP_008770463.1 | 3 | 372 | |||
| XM_008772242.1 | XP_008770464.1 | 3 | 321 | |||
| GenBank mRNA | BC169107.1 | - | 3 | 377 | 7130835 | |

Cmtm3
Genomic Map