siRNA Details
| Gene Symbol: Areg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017123.1 | NP_058819.1 | 5 | 893 | ||
| GenBank mRNA | X55183.1 | - | 5 | 893 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Areg| Gene Symbol: Areg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017123.1 | NP_058819.1 | 5 | 893 | ||
| GenBank mRNA | X55183.1 | - | 5 | 893 | ||