siRNA Details
| Gene Symbol: Zfp319 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001170478.1 | NP_001163949.1 | 2 | 1221 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp319| Gene Symbol: Zfp319 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001170478.1 | NP_001163949.1 | 2 | 1221 | ||