siRNA Details
| Gene Symbol: Pgrmc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021766.1 | NP_068534.1 | 3 | 700 | ||
| GenBank mRNA | AF163321.1 | - | 3 | 658 | ||
| BC062073.1 | - | 3 | 661 | 5599449 | ||
| U63315.1 | - | 3 | 700 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pgrmc1| Gene Symbol: Pgrmc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021766.1 | NP_068534.1 | 3 | 700 | ||
| GenBank mRNA | AF163321.1 | - | 3 | 658 | ||
| BC062073.1 | - | 3 | 661 | 5599449 | ||
| U63315.1 | - | 3 | 700 | |||