siRNA Details
| Gene Symbol: Hsf4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106177.1 | NP_001099647.1 | 12 | 1237 | ||
| XM_006255453.2 | XP_006255515.1 | 13 | 1895 | |||
| XM_006255454.2 | XP_006255516.1 | 13 | 1805 | |||
| XM_006255455.2 | XP_006255517.1 | 12 | 1786 | |||

Hsf4
Genomic Map