siRNA Details
| Gene Symbol: Tppp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009639.1 | NP_001009639.1 | 4 | 614 | ||
| XM_006255464.2 | XP_006255526.1 | 4 | 644 | |||
| GenBank mRNA | BC087585.1 | - | 4 | 614 | 7317980 | |
| FQ213895.1 | - | 3 | 756 | |||
| FQ214516.1 | - | 4 | 628 | |||
| FQ214740.1 | - | 4 | 618 | |||
| FQ216797.1 | - | 4 | 639 | |||
| FQ217207.1 | - | 4 | 615 | |||

Tppp3
Genomic Map