siRNA Details
| Gene Symbol: Il18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019165.1 | NP_062038.1 | 3 | 141 | ||
| GenBank mRNA | AY258448.1 | - | 3 | 141 | ||
| U77776.1 | - | 3 | 141 | |||
| U77777.1 | - | 3 | 141 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Il18| Gene Symbol: Il18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019165.1 | NP_062038.1 | 3 | 141 | ||
| GenBank mRNA | AY258448.1 | - | 3 | 141 | ||
| U77776.1 | - | 3 | 141 | |||
| U77777.1 | - | 3 | 141 | |||