siRNA Details
| Gene Symbol: Cirh1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009640.1 | NP_001009640.1 | 2 | 148 | ||
| XM_006255562.1 | XP_006255624.1 | 2 | 194 | |||
| XM_006255563.2 | XP_006255625.1 | 2 | 357 | |||
| XM_008772488.1 | XP_008770710.1 | 2 | 147 | |||
| GenBank mRNA | BC088461.1 | - | 2 | 148 | 7127580 | |

Cirh1a
Genomic Map