siRNA Details
| Gene Symbol: Ddx19a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005381.2 | NP_001005381.2 | 1, 2 | 133 | ||
| GenBank mRNA | BC079094.1 | - | 1, 2 | 104 | 7123989 | |
| CB582878.1 | - | 1, 2 | 133 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ddx19a| Gene Symbol: Ddx19a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005381.2 | NP_001005381.2 | 1, 2 | 133 | ||
| GenBank mRNA | BC079094.1 | - | 1, 2 | 104 | 7123989 | |
| CB582878.1 | - | 1, 2 | 133 | |||