siRNA Details
| Gene Symbol: Ddx19a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005381.2 | NP_001005381.2 | 5 | 403 | ||
| XM_008772490.1 | XP_008770712.1 | 189 | ||||
| GenBank mRNA | BC079094.1 | - | 5 | 374 | 7123989 | |
| CB582878.1 | - | 5 | 403 | |||

Ddx19a
Genomic Map