siRNA Details
| Gene Symbol: Syngr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019166.2 | NP_062039.1 | 2 | 367 | ||
| XM_006241971.2 | XP_006242033.1 | 2 | 372 | |||
| GenBank mRNA | DN932005.1 | - | 2 | 329 | ||
| DV724604.1 | - | 1 | 173 | |||
| U39549.1 | - | 2 | 336 | |||

Syngr1
Genomic Map